Thiophene-2-carboxamide, 99%
Thiophene-2-carboxamide, 99%
Thiophene-2-carboxamide, 99%
Thiophene-2-carboxamide, 99%
Thermo Scientific Chemicals

Thiophene-2-carboxamide, 99%

CAS: 5813-89-8 | C5H5NOS | 127.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15219.14
also known as A15219-14
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS5813-89-8
IUPAC Namethiophene-2-carboxamide
Molecular FormulaC5H5NOS
InChI KeyDENPQNAWGQXKCU-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)175.0-181.0?C
Appearance (Color)Pale cream to pale brown
Assay (HPLC)≥98.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only