(4-Bromobenzyl)triphenylphosphonium bromide, 98%
(4-Bromobenzyl)triphenylphosphonium bromide, 98%
(4-Bromobenzyl)triphenylphosphonium bromide, 98%
(4-Bromobenzyl)triphenylphosphonium bromide, 98%
Thermo Scientific Chemicals

(4-Bromobenzyl)triphenylphosphonium bromide, 98%

CAS: 51044-13-4 | C25H21Br2P | 512.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15220.14
also known as A15220-14
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS51044-13-4
IUPAC Name[(4-bromophenyl)methyl]triphenylphosphanium bromide
Molecular FormulaC25H21Br2P
InChI KeyFQJYKXVQABPCRA-UHFFFAOYSA-M
SMILES[Br-].BrC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
Melting Point (clear melt)272.0-279.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only