4-Nitrobenzyl bromide, 97+%
4-Nitrobenzyl bromide, 97+%
4-Nitrobenzyl bromide, 97+%
4-Nitrobenzyl bromide, 97+%
Thermo Scientific Chemicals

4-Nitrobenzyl bromide, 97+%

CAS: 100-11-8 | C7H6BrNO2 | 216.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15236.22
also known as A15236-22
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS100-11-8
IUPAC Name1-(bromomethyl)-4-nitrobenzene
Molecular FormulaC7H6BrNO2
InChI KeyVOLRSQPSJGXRNJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)>97.0%
Solution Test5% in acetone: clear - no visible particlulates
FormCrystalline powder
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

Solubility
It hydrolyzes in water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
RUO – Research Use Only

General References:

  1. Josef Prousek. The course of reaction of 4-nitrobenzyl bromide and 5-nitrofurfuryl bromide with bases: Operation of anion-radical mechanism. Collect. Czech. Chem. Commun. 1982, 47,(5), 1334-1338.
  2. Paul G.Bulger; Ian F.Cottrell; Cameron J.Cowden; Antony J.Davies; Ulf-H Dolling. An investigation into the alkylation of 1,2,4-triazole. Tetrahedron Letters. 2000, 41,(8), 1297-1301.
  3. Has been used as a protecting reagent for the carboxyl group in peptide synthesis: Helv. Chim. Acta, 42, 972 (1959); see 4-Nitrobenzyl chloride, A15749, and Appendix 6. Has also been used in ß-lactam synthesis. Cleavage by sodium sulfide avoids hydrogenolysis problems otherwise encountered with these sulfur-containing nuclei: J. Org. Chem., 43, 1243 (1978).
  4. For the protection of alcohols, introduction of the group by the use of silver oxide has been recommended. Selective cleavage can be accomplished with DDQ (2,3-Dichloro-5,6-dicyanobenzoquinone, A11879) or by anodic oxidation. Many other benzyl-type protecting groups are unaffected by these conditions: Tetrahedron Lett., 31, 389 (1990). Indium metal in the presence of ammonium chloride is an effective deprotection system for 4-nitrobenzyl ethers and esters: J. Chem. Soc., Perkin 1, 955 (2001).