2-Phenylethanol, 98+%
2-Phenylethanol, 98+%
2-Phenylethanol, 98+%
Thermo Scientific Chemicals

2-Phenylethanol, 98+%

CAS: 60-12-8 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A15241.0B
also known as A15241-0B
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS60-12-8
IUPAC Name2-phenylethan-1-ol
Molecular FormulaC8H10O
InChI KeyWRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.5290-1.5350 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

Solubility
Miscible with chloroform.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Etschmann, M.; Bluemke, W.; Sell, D.; Schrader, J. Biotechnological production of 2-phenylethanol. Applied Microbiology and Biotechnology. 2002, 59, (1), 1-8.
  2. Kenji Hanabusa.; Yukinobu Maesaka.; Mutsumi Kimura.; Hirofusa Shirai. New gelators based on 2-amino-2-phenylethanol: Close gelator-chiral structure relationship. Tetrahedron Letters. 1999, 40, (12), 2385-2388.