6-Methyl-1,2,3,4-tetrahydroquinoline, 98%
6-Methyl-1,2,3,4-tetrahydroquinoline, 98%
6-Methyl-1,2,3,4-tetrahydroquinoline, 98%
6-Methyl-1,2,3,4-tetrahydroquinoline, 98%
Thermo Scientific Chemicals

6-Methyl-1,2,3,4-tetrahydroquinoline, 98%

CAS: 91-61-2 | C10H13N | 147.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A15245.18
also known as A15245-18
Price (USD)
661.65
Online Exclusive
735.00
Save 73.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
661.65
Online Exclusive
735.00
Save 73.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS91-61-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale brown or clear colorless to yellow to pale brown
FormFused solid or liquid as melt
Assay (GC)≥97.5%
Melting Point (clear melt)32.0-38.0?C
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. V. Krishnakumar; N.Prabavathi; S.Muthunatesan. Density functional theory calculations and vibrational spectra of 6-methyl 1,2,3,4-tetrahyroquinoline. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2008, 69,(3), 853-859.
  2. Abbas Alvanipour; Lowell D.Kispert. Cobalt stearate-aluminum alkyl catalyzed hydrogenation of substituted quinolines, isoquinoline and naphthalene. Journal of Molecular Catalysis. 1988, 48,(2-3), 277-283.