Linoleic acid, tech. 75%
Linoleic acid, tech. 75%
Linoleic acid, tech. 75%
Thermo Scientific Chemicals

Linoleic acid, tech. 75%

CAS: 60-33-3 | C18H32O2 | 280.452 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15265.09
also known as A15265-09
Price (USD)
51.65
Online Exclusive
57.40
Save 5.75 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS60-33-3
IUPAC Name(9Z,12Z)-octadeca-9,12-dienoic acid
Molecular FormulaC18H32O2
InChI KeyOYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILESCCCCCC=C/CC=C/CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥70.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only