2,2'-Furoin, 98%
2,2'-Furoin, 98%
2,2'-Furoin, 98%
Thermo Scientific Chemicals

2,2'-Furoin, 98%

CAS: 552-86-3 | C10H8O4 | 192.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15272.22
also known as A15272-22
Price (USD)
779.65
Special offer
Online exclusive
Ends: 30-Jun-2026
917.00
Save 137.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
779.65
Special offer
Online exclusive
Ends: 30-Jun-2026
917.00
Save 137.35 (15%)
Each
Chemical Identifiers
CAS552-86-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown or yellow-orange
FormPowder, crystals, crystalline powder and/or chunks
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)133-142?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only