5-Methylcyclohexane-1,3-dione, 98%
5-Methylcyclohexane-1,3-dione, 98%
5-Methylcyclohexane-1,3-dione, 98%
Thermo Scientific Chemicals

5-Methylcyclohexane-1,3-dione, 98%

CAS: 4341-24-6 | C7H10O2 | 126.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A15280.06
also known as A15280-06
Price (USD)
110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
130.00
Save 19.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
130.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS4341-24-6
IUPAC Name(5S)-3-hydroxy-5-methylcyclohex-2-en-1-one
Molecular FormulaC7H10O2
InChI KeyMNOUUYHZNVGKSQ-YFKPBYRVSA-N
SMILESC[C@H]1CC(O)=CC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to orange
FormCrystals or powder or crystalline powder
Melting Point (clear melt)123.0-130.0°C
Assay (GC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only