(4-Nitrobenzyl)triphenylphosphonium bromide, 98%
(4-Nitrobenzyl)triphenylphosphonium bromide, 98%
(4-Nitrobenzyl)triphenylphosphonium bromide, 98%
Thermo Scientific Chemicals

(4-Nitrobenzyl)triphenylphosphonium bromide, 98%

CAS: 2767-70-6 | C25H21BrNO2P | 478.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15288.06
also known as A15288-06
Price (USD)
26.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
26.60
Each
Chemical Identifiers
CAS2767-70-6
IUPAC Name[(4-nitrophenyl)methyl]triphenylphosphanium bromide
Molecular FormulaC25H21BrNO2P
InChI KeyIPJPTPFIJLFWLP-UHFFFAOYSA-M
SMILES[Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only