3,5-Bis(trifluoromethyl)aniline, 98%
3,5-Bis(trifluoromethyl)aniline, 98%
3,5-Bis(trifluoromethyl)aniline, 98%
3,5-Bis(trifluoromethyl)aniline, 98%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)aniline, 98%

CAS: 328-74-5 | C8H5F6N | 229.125 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A15289.09
also known as A15289-09
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.40
Save 3.75 (13%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.40
Save 3.75 (13%)
Each
Add to cart
Chemical Identifiers
CAS328-74-5
IUPAC Name3,5-bis(trifluoromethyl)aniline
Molecular FormulaC8H5F6N
InChI KeyCDIDGWDGQGVCIB-UHFFFAOYSA-N
SMILESNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to yellow-brown or pale brown
Refractive Index1.4320-1.4360 @ 20?C
FormLiquid
Assay (GC)≥97.5%
3,5-Bis(trifluoromethyl)aniline was used in the synthesis of N-[2-hydroxy-1-naphthylidene]-3, 5-bis(trifluoromethyl)aniline, Schiff?s base and 5,7-bis(trifluoromethyl)aniline. It was also used in the synthesis of N-1-phenylethyl-3,5-bis(trifluoromethyl)aniline via titanium-catalyzed hydroamination reaction and N,N?-bis[3,5-bis(tri-fluoromethyl)phenyl]thiourea, organocatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)aniline was used in the synthesis of N-[2-hydroxy-1-naphthylidene]-3, 5-bis(trifluoromethyl)aniline, Schiff′s base and 5,7-bis(trifluoromethyl)aniline. It was also used in the synthesis of N-1-phenylethyl-3,5-bis(trifluoromethyl)aniline via titanium-catalyzed hydroamination reaction and N,N′-bis[3,5-bis(tri-fluoromethyl)phenyl]thiourea, organocatalyst.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ludwig T.Kaspar; Benjamin Fingerhut; Lutz Ackermann. Titanium-catalyzed intermolecular hydroamination of vinylarene. Angewandte Chemie (International Edition). 2005, 44,(37), 5972-5974.
  2. Yunoh Jung; Kang-Jun Baeg; Dong-Yu Kim; Takao Someya; Soo Young Park. A thermally resistant and air-stable n-type organic semiconductor: Naphthalene diimide of 3,5-bis-trifluoromethyl aniline. Synthetic Metals. 2009, 159,(19-20), 2117-2121.