3-Nitrophthalimide, 98%
3-Nitrophthalimide, 98%
3-Nitrophthalimide, 98%
3-Nitrophthalimide, 98%
Thermo Scientific Chemicals

3-Nitrophthalimide, 98%

CAS: 603-62-3 | C8H4N2O4 | 192.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15291.22
also known as A15291-22
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS603-62-3
IUPAC Name4-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC8H4N2O4
InChI KeyBONIIQYTWOPUQI-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC2=C1C(=O)NC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)215.0-222.0?C
3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Kalyan K. Sethia; Saurabh M. Vermaa; Muhammet Tanc; Gaultier Purper; Gaetan Calafato; Fabrizio Carta; Claudiu T. Supuran. Carbonic anhydrase inhibitors: Synthesis and inhibition of the human carbonic anhydrase isoforms I, II, IX and XII with benzene sulfonamides incorporating 4- and 3-nitrophthalimide moieties.Bioorganic & Medicinal Chemistry.2014, 22 1586-1595.
  2. C. Glidewell; J. N. Low; J. M. S. Skakle and J. L. Wardell. Isomeric nitrophthalimides: sheets built from N-H...O and C-H...O hydrogen bonds.Crystal Structure Communications.2004, 60 872-875 .