2-Bromonaphthalene, 98+%
2-Bromonaphthalene, 98+%
2-Bromonaphthalene, 98+%
Thermo Scientific Chemicals

2-Bromonaphthalene, 98+%

CAS: 580-13-2 | C10H7Br | 207.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15300.14
also known as A15300-14
Price (USD)
143.65
Special offer
Online exclusive
Ends: 30-Jun-2026
169.00
Save 25.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
143.65
Special offer
Online exclusive
Ends: 30-Jun-2026
169.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS580-13-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or lumps
Assay (GC)≥98.0%
2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. The Grignard reagent has been reacted with N-benzylproline methyl ester in the synthesis of oxazaborolidines (compare (S)-2-Methyl-CBS-oxazaborolidine monohydrate, L09219) for use as chiral auxiliaries: J. Org. Chem., 56, 442 (1991).
  2. Hopkins, B. A.; Garlets, Z. J.; Wolfe, J. P. Development of Enantioselective Palladium-Catalyzed Alkene Carboalkoxylation Reactions for the Synthesis of Tetrahydrofurans. Angew. Chem. Int. Ed. 2015, 54 (45), 13390-13392
  3. Xu, L.; Suo, H.; Liang, X.; Wang, L.; Guo, Y.; Jiang, S. Au nanoparticle decorated graphene oxide as a novel coating for solid-phase microextraction. RSC Adv. 2015, 5 (52), 41536-41543.