3-Nitrophenol, 98+%
3-Nitrophenol, 98+%
3-Nitrophenol, 98+%
Thermo Scientific Chemicals

3-Nitrophenol, 98+%

CAS: 554-84-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15312.09
also known as A15312-09
Price (USD)
68.65
Online exclusive
75.90
Save 7.25
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
68.65
Online exclusive
75.90
Save 7.25
Each
Chemical Identifiers
CAS554-84-7
IUPAC Name3-nitrophenol
Molecular FormulaC6H5NO3
InChI KeyRTZZCYNQPHTPPL-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to orange to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)93.0-99.0?C

3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).

Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

General References:

  1. A. Alif; P. Boule; J. Lemaire. Photochemistry and environment XII: Phototransformation of 3-nitrophenol in aqueous solution.Journal of Photochemistry and Photobiology A.1990, 50 331-342.
  2. Mohammad R.Haghighi Podeh; Sanjoy K. Bhattacharya; Mingbo Qu. Effects of nitrophenols on acetate utilizing methanogenic systems.Water Research.1995, 29 391-399.
  3. Acid-base indicator: approx pH 6.6 - 8.6.
  4. Has been used for the formation of active esters. These are more stable than the corresponding 4-nitrophenyl esters, but are sufficiently reactive for use in peptide synthesis: Synthesis, 465 (1979).