Nicotinoyl chloride hydrochloride, 96%
Nicotinoyl chloride hydrochloride, 96%
Nicotinoyl chloride hydrochloride, 96%
Nicotinoyl chloride hydrochloride, 96%
Thermo Scientific Chemicals

Nicotinoyl chloride hydrochloride, 96%

CAS: 20260-53-1 | C6H5Cl2NO | 178.012 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15317.18
also known as A15317-18
Price (USD)
91.65
Special offer
Online exclusive
Ends: 30-Jun-2026
108.00
Save 16.35 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
91.65
Special offer
Online exclusive
Ends: 30-Jun-2026
108.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS20260-53-1
IUPAC Namehydrogen pyridine-3-carbonyl chloride chloride
Molecular FormulaC6H5Cl2NO
InChI KeyMSYBLBLAMDYKKZ-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to gray
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. K.W.Morin; L.I.Wiebe; E.E.Knaus. Synthesis of brain-targeted 1-(2-deoxy-2-fluoro-beta-D-ribofuranosyl)-(E)-5-(2-iodovinyl)uracil coupled to a dihydropyridine <---> pyridinium salt redox chemical-delivery system. Carbohydrate Research. 1993, 249,(1), 109-116.
  2. K.Sakata; M.Itoh. Synthesis and characterization of 7,16-dinicotinoyl- and 7,16-diisonico-tinoyltetraaza[14]annulene and their nickel(II) and copper(II) complexes. Journal of heterocyclic chemistry. 1992, 29,(4), 921-926.