4-Bromophenylacetic acid, 99%
4-Bromophenylacetic acid, 99%
4-Bromophenylacetic acid, 99%
4-Bromophenylacetic acid, 99%
Thermo Scientific Chemicals

4-Bromophenylacetic acid, 99%

CAS: 1878-68-8 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A15325.22
also known as A15325-22
Price (USD)
153.65
Special offer
Online exclusive
Ends: 31-Dec-2025
181.00
Save 27.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
153.65
Special offer
Online exclusive
Ends: 31-Dec-2025
181.00
Save 27.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1878-68-8
IUPAC Name2-(4-bromophenyl)acetic acid
Molecular FormulaC8H7BrO2
InChI KeyQOWSWEBLNVACCL-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Melting Point (clear melt)113.0-120.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only