2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
Thermo Scientific Chemicals

2-Pyrrolidinone, 99%

CAS: 616-45-5 | C4H7NO | 85.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number A15332.A3
also known as A15332-A3
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
147.00
Save 22.35 (15%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
147.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS616-45-5
IUPAC Namepyrrolidin-2-one
Molecular FormulaC4H7NO
InChI KeyHNJBEVLQSNELDL-UHFFFAOYSA-N
SMILESO=C1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.5%
Water Content (Karl Fischer Titration)≤0.5%
Refractive Index1.4860-1.4890 @ 20?C
Appearance (Color)Clear colorless to pale yellow
View more
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

Solubility
Miscible with water and other organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only

General References:

  1. Jiang, H.; Yuana, H.; Liang, Y.; Xia, S.; Zhao, Q.; Wu, Q.; Zhang, L.; Lianga, Z.; Zhang, Y. A hydrophilic immobilized trypsin reactor with N-vinyl-2-pyrrolidinone modified polymer microparticles as matrix for highly efficient protein digestion with low peptide residue. J. Chromatogr. A 2012, 1246, 111-116.
  2. Shahbuddin, M.; Bullock, A. J.; MacNeil, S.; Rimme, S. Glucomannan-poly(N-vinyl pyrrolidinone) bicomponent hydrogels for wound healing. J. Mater. Chem. B 2014, 2 (6), 727-738.