Thermo Scientific Chemicals

m-Phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A15339.30
250 g, Each
Thermo Scientific Chemicals

m-Phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A15339.30
250 g, Each
Quantity
Catalog number: A15339.30
also known as A15339-30
Price (USD)
Price: 36.80
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-45-2
IUPAC Name
benzene-1,3-diamine
Molecular Formula
C6H8N2
InChI Key
WZCQRUWWHSTZEM-UHFFFAOYSA-N
SMILES
NC1=CC(N)=CC=C1
Appearance (Color)
White to dark brown or grey
Form
Crystalline powder or flakes or granular or pelleted solid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
60-68?C

Description

m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.

Solubility
Soluble in water, methanol, acetone, DMF, MEK, dioxane

Notes
Air & Light Sensitive. Store in the dark. Store under dry argon. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text