Dicyclohexano-18-crown-6, mixture of isomers, 97%
Dicyclohexano-18-crown-6, mixture of isomers, 97%
Dicyclohexano-18-crown-6, mixture of isomers, 97%
Thermo Scientific Chemicals

Dicyclohexano-18-crown-6, mixture of isomers, 97%

CAS: 16069-36-6 | C20H36O6 | 372.502 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
25 g
Catalog number A15344.03
also known as A15344-03
Price (USD)
30.65
Online Exclusive
33.80
Save 3.15 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
33.80
Save 3.15 (9%)
Each
Chemical Identifiers
CAS16069-36-6
IUPAC Name2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosane
Molecular FormulaC20H36O6
InChI KeyBBGKDYHZQOSNMU-UHFFFAOYNA-N
SMILESC1CCC2OCCOCCOC3CCCCC3OCCOCCOC2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Vinod K Gupta; Sudeshna Chandr; Rajni Mangla. Dicyclohexano-18-crown-6 as active material in PVC matrix membrane for the fabrication of cadmium selective potentiometric sensor. Electrochimica Acta.2002, 47 1579-1586.
  2. E. P. Horwitza; M. L. Dietza & D. E. Fishera. Extraction of stoontium from nitric acid solutions using dicyclohexano-18-crown-5 and its derivatives. Electrochimica Acta.1990, 8 557-572.
  3. Crown ether (see Appendix 2). Increases the rate of acetolysis of benzyl chloride in acetonitrile by a factor of >400: Synthesis, 117 (1981).
  4. In combination with potassium permanganate, effects the stereoselective cis-hydroxylation of a double bond from the less-hindered side: Chem. Lett., 173 (1983).