Methyl 3-amino-4-methylbenzoate, 97%
Methyl 3-amino-4-methylbenzoate, 97%
Methyl 3-amino-4-methylbenzoate, 97%
Thermo Scientific Chemicals

Methyl 3-amino-4-methylbenzoate, 97%

CAS: 18595-18-1 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15348.06
also known as A15348-06
Price (USD)
26.65
Online exclusive
29.80
Save 3.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
26.65
Online exclusive
29.80
Save 3.15
Each
Chemical Identifiers
CAS18595-18-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)110.0-119.0?C
Appearance (Color)White to cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only