2-Acetyl-1-methylpyrrole, 98%
2-Acetyl-1-methylpyrrole, 98%
2-Acetyl-1-methylpyrrole, 98%
Thermo Scientific Chemicals

2-Acetyl-1-methylpyrrole, 98%

CAS: 932-16-1 | C7H9NO | 123.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15353.14
also known as A15353-14
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS932-16-1
IUPAC Name1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one
Molecular FormulaC7H9NO
InChI KeyNZFLWVDXYUGFAV-UHFFFAOYSA-N
SMILESCN1C=CC=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥97.5%
Refractive Index1.5390-1.5430 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only