2,5-Diphenyl-p-benzoquinone, 96%
2,5-Diphenyl-p-benzoquinone, 96%
2,5-Diphenyl-p-benzoquinone, 96%
Thermo Scientific Chemicals

2,5-Diphenyl-p-benzoquinone, 96%

CAS: 844-51-9 | C18H12O2 | 260.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15369.14
also known as A15369-14
Price (USD)
145.65
Special offer
Online exclusive
Ends: 30-Jun-2026
171.00
Save 25.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
145.65
Special offer
Online exclusive
Ends: 30-Jun-2026
171.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS844-51-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥95.0%
Melting Point (clear melt)213-221°C
Appearance (Color)Yellow to orange
FormPowder
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

Solubility
Soluble in hot Toluene (almost transparency).

Notes
Incompatible with Oxidizing agents, Reducing agent.
RUO – Research Use Only