2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate
2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate
2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate
Thermo Scientific Chemicals

2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate

CAS: 6482-24-2 | C3H7BrO | 138.992 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number A15370.14
also known as A15370-14
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.60
Save 10.95 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.60
Save 10.95 (15%)
Each
Chemical Identifiers
CAS6482-24-2
IUPAC Name1-bromo-2-methoxyethane
Molecular FormulaC3H7BrO
InChI KeyYZUPZGFPHUVJKC-UHFFFAOYSA-N
SMILESCOCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Refractive Index1.4450-1.4510 @ 20°C
Appearance (Color)Clear colourless to yellow
FormLiquid
2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Solubility
Soluble in water.14,4 g/L at 25°C

Notes
Protect from heat. Incompatible with heat, strong bases and oxidizing agents. Store at 2-8°C.
RUO – Research Use Only

General References:

  1. Ralph C. Tallman. Reactions of 2-Halogen Ethers. I. The Action of Magnesium on Alkyl 2-Bromo-ethyl Ethers.J. Am. Chem. Soc.1934, 56 (1), 126-129.
  2. Hiroatsu Matsuura; Nobuyuki Miyauchi; Hiromu Murata; Masaaki Sakakibara. Vibrational Spectra and Rotational Isomerism of 2-Chloro-and 2-Bromoethyl Methyl Sulfides. Bulletin of the Chemical Society of Japan.1979, 52 344-350.