Tri(m-tolyl)phosphine, 98+%
Tri(m-tolyl)phosphine, 98+%
Tri(m-tolyl)phosphine, 98+%
Thermo Scientific Chemicals

Tri(m-tolyl)phosphine, 98+%

CAS: 6224-63-1 | C21H21P | 304.373 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15372.06
also known as A15372-06
Price (USD)
56.65
Online Exclusive
62.80
Save 6.15 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS6224-63-1
IUPAC Nametris(3-methylphenyl)phosphane
Molecular FormulaC21H21P
InChI KeyLFNXCUNDYSYVJY-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1)P(C1=CC(C)=CC=C1)C1=CC=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormPowder
Melting Point (clear melt)98.0-105.0?C
Appearance (Color)White
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only
T. S. Cameron; K. D. Howlett and K. Miller. Investigation of the phosphorus-carbon bond lengths in aromatic phosphines. II. Crystal and molecular structures of tri-m-tolylphosphine, tri-m-tolylphosphine sulphide and tri-m-tolylphosphine selenide.Acta Cryst.1978, 34 1639-1644.