1,4-Diethoxybenzene, 98%
1,4-Diethoxybenzene, 98%
1,4-Diethoxybenzene, 98%
Thermo Scientific Chemicals

1,4-Diethoxybenzene, 98%

CAS: 122-95-2 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15385.22
also known as A15385-22
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
127.00
Save 19.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
127.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS122-95-2
IUPAC Name1,4-diethoxybenzene
Molecular FormulaC10H14O2
InChI KeyVWGNFIQXBYRDCH-UHFFFAOYSA-N
SMILESCCOC1=CC=C(OCC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)69.0-75.0?C
Appearance (Color)White
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused/lumpy solid or chunks
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only