2-Chloropyrimidine, 98%
2-Chloropyrimidine, 98%
2-Chloropyrimidine, 98%
Thermo Scientific Chemicals

2-Chloropyrimidine, 98%

CAS: 1722-12-9 | C4H3ClN2 | 114.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number A15394.09
also known as A15394-09
Price (USD)
63.65
Online Exclusive
70.00
Save 6.35 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.00
Save 6.35 (9%)
Each
Chemical Identifiers
CAS1722-12-9
IUPAC Name2-chloropyrimidine
Molecular FormulaC4H3ClN2
InChI KeyUNCQVRBWJWWJBF-UHFFFAOYSA-N
SMILESClC1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown or pale yellow to yellow to pale orange
Water Content (Karl Fischer Titration)≤1.0%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
View more
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4?-(1,1?-(5-(2-methoxyphenoxy)-[2,2?-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

Solubility
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
RUO – Research Use Only

General References:

  1. Jeanne-Marie Bégouin and Corinne Gosmini. Cobalt-Catalyzed Cross-Coupling Between In Situ Prepared Arylzinc Halides and 2-Chloropyrimidine or 2-Chloropyrazine. J. Org. Chem. 2009, 74 (8), 3221-3224.
  2. Y.Anantharama Sarma. Planar vibrations of 2-chloropyrimidine. Spectrochimica Acta Part A: Molecular Spectroscopy. 1974, 30 (9), 1801-1806.
  3. Alkoxylation with 3-butyn-1-ol, followed by intramolecular Diels-Alder reaction, leads to a fused pyridine system, by a sequence analogous to that shown for 2-Chloropyrazine, A10108: Tetrahedron, 45, 803 (1989). For a similar sequence using the anion of an alkynylmalononitrile, see: Tetrahedron, 45, 5151 (1989). For synthesis of a benzofuropyridine by the cyclization of a 2-(alkynylphenoxy)pyrimidine, see: Tetrahedron, 45, 6511 (1989).
  4. Organolithium reagents add to the 1,6-imine bond, giving, after aromatization with DDQ, 6-substituted 2-chloropyrimidines: J. Org. Chem., 53, 4137 (1988).