Search
View additional product information for Aluminon - FAQs (A15401.30, A15401.09, A15401.18)
1 product FAQs found
Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.