1-Chloro-2-nitrobenzene, 99%
1-Chloro-2-nitrobenzene, 99%
1-Chloro-2-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Chloro-2-nitrobenzene, 99%

CAS: 88-73-3 | C6H4ClNO2 | 157.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15403.0B
also known as A15403-0B
Price (USD)
70.65
Online Exclusive
77.80
Save 7.15 (9%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
77.80
Save 7.15 (9%)
Each
Chemical Identifiers
CAS88-73-3
IUPAC Name1-chloro-2-nitrobenzene
Molecular FormulaC6H4ClNO2
InChI KeyBFCFYVKQTRLZHA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Yellow
Melting Point (clear melt)30.0-36.0?C
FormCrystals or powder or crystalline powder or fused solid
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

Solubility
Soluble in water 0.59 g/L.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. I. Mossakowska and G. Wójcik. 1-Chloro-2-nitro­benzene: N-O...Cl halogen bonds and aromatic [pi]-[pi] stacking, and thermal vibrations in the vicinity of the melting point. Acta Cryst. 2007, c63, o123-o125.
  2. Robert Gatermann; Heinrich Hühnerfuss; Gerhard Rimkus; Manfred Wolf; Stephan Franke. The distribution of nitrobenzene and other nitroaromatic compounds in the North Sea. Marine Pollution Bulletin. 1995, 30(3), 221-227.