4-n-Heptylphenol, 98+%
4-n-Heptylphenol, 98+%
4-n-Heptylphenol, 98+%
Thermo Scientific Chemicals

4-n-Heptylphenol, 98+%

CAS: 1987-50-4 | C13H20O | 192.302 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15407.03
also known as A15407-03
Price (USD)
43.65
Online exclusive
48.30
Save 4.65
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.65
Online exclusive
48.30
Save 4.65
Each
Chemical Identifiers
CAS1987-50-4
IUPAC Name4-heptylphenol
Molecular FormulaC13H20O
InChI KeyKNDDEFBFJLKPFE-UHFFFAOYSA-N
SMILESCCCCCCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormFused solid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)26.0-32.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only