2-Naphthoxyacetic acid, 97%
2-Naphthoxyacetic acid, 97%
2-Naphthoxyacetic acid, 97%
2-Naphthoxyacetic acid, 97%
Thermo Scientific Chemicals

2-Naphthoxyacetic acid, 97%

CAS: 120-23-0 | C12H10O3 | 202.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15412.22
also known as A15412-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS120-23-0
IUPAC Name2-(naphthalen-2-yloxy)acetic acid
Molecular FormulaC12H10O3
InChI KeyRZCJYMOBWVJQGV-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey or brown / pink
Assay (Aqueous acid-base Titration)>96.0%
Formpowder
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.

Notes
Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Tsilikochrisos, G.; Tsaniklidis, G.; Delis, C.; Nikoloudakis, N.; Aivalakis, G. Glutamate dehydrogenase is differentially regulated in seeded and parthenocarpic tomato fruits during crop development and postharvest storage. Sci. Hortic. 2015, 181, 34-42.
  2. Song, X. Y.; Chen, J.; Shi, Y. P. Electromembrane extraction based on carbon nanotubes reinforced hollow fiber for the determination of plant hormones. New J. Chem. 2015, 39 (12), 9191-9199.