3,5-Dimethylisoxazole-4-carboxylic acid, 99%
3,5-Dimethylisoxazole-4-carboxylic acid, 99%
3,5-Dimethylisoxazole-4-carboxylic acid, 99%
Thermo Scientific Chemicals

3,5-Dimethylisoxazole-4-carboxylic acid, 99%

CAS: 2510-36-3 | C6H7NO3 | 141.126 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15419.14
also known as A15419-14
Price (USD)
437.65
Online Exclusive
486.00
Save 48.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
437.65
Online Exclusive
486.00
Save 48.35 (10%)
Each
Chemical Identifiers
CAS2510-36-3
IUPAC Namedimethyl-1,2-oxazole-4-carboxylate
Molecular FormulaC6H6NO3
InChI KeyIJEUISLJVBUNRE-UHFFFAOYSA-M
SMILESCC1=C(C([O-])=O)C(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown
Assay (Aqueous acid-base Titration)>97.5%
Formpowder
Melting Point139-144?C (with ≤3?C range)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only