4-Chloro-m-phenylenediamine, 97%
4-Chloro-m-phenylenediamine, 97%
4-Chloro-m-phenylenediamine, 97%
Thermo Scientific Chemicals

4-Chloro-m-phenylenediamine, 97%

CAS: 5131-60-2 | C6H7ClN2 | 142.586 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15430.06
also known as A15430-06
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.70
Save 11.05 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.70
Save 11.05 (15%)
Each
Chemical Identifiers
CAS5131-60-2
IUPAC Name4-chlorobenzene-1,3-diamine
Molecular FormulaC6H7ClN2
InChI KeyZWUBBMDHSZDNTA-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey to red to brown
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)83.5-92.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only