2,4-Dinitrobenzaldehyde, 98%
2,4-Dinitrobenzaldehyde, 98%
2,4-Dinitrobenzaldehyde, 98%
Thermo Scientific Chemicals

2,4-Dinitrobenzaldehyde, 98%

CAS: 528-75-6 | C7H4N2O5 | 196.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15452.06
also known as A15452-06
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS528-75-6
IUPAC Name2,4-dinitrobenzaldehyde
Molecular FormulaC7H4N2O5
InChI KeyZILXIZUBLXVYPI-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=O)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Free acid (titration)≤1.0%
Identification (FTIR)Conforms
Melting Point64.5-70.5°C
Appearance (Color)Cream or yellow or pale orange to orange to orange-brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only