4-Fluoro-3-methylaniline, 97%
4-Fluoro-3-methylaniline, 97%
4-Fluoro-3-methylaniline, 97%
Thermo Scientific Chemicals

4-Fluoro-3-methylaniline, 97%

CAS: 452-69-7 | C7H8FN | 125.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15482.14
also known as A15482-14
Price (USD)
171.65
Online Exclusive
190.00
Save 18.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
171.65
Online Exclusive
190.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS452-69-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown or purple
FormLiquid or solid or crystals
Assay (GC)≥96.0%
Melting point32-38°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only