2-Chloro-4-fluoroaniline, 97%
2-Chloro-4-fluoroaniline, 97%
2-Chloro-4-fluoroaniline, 97%
Thermo Scientific Chemicals

2-Chloro-4-fluoroaniline, 97%

CAS: 2106-02-7 | C6H5ClFN | 145.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15492.14
also known as A15492-14
Price (USD)
127.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
127.00
Each
Chemical Identifiers
CAS2106-02-7
IUPAC Name2-chloro-4-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyXRAKCYJTJGTSMM-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5515-1.5565 @ 20°C
Appearance (Color)Clear colorless to yellow to brown or pale pink to dark purple
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only