1-Chloro-2,4-difluorobenzene, 98%
1-Chloro-2,4-difluorobenzene, 98%
1-Chloro-2,4-difluorobenzene, 98%
Thermo Scientific Chemicals

1-Chloro-2,4-difluorobenzene, 98%

CAS: 1435-44-5 | C6H3ClF2 | 148.537 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15495.30
also known as A15495-30
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS1435-44-5
IUPAC Name1-chloro-2,4-difluorobenzene
Molecular FormulaC6H3ClF2
InChI KeyAJCSNHQKXUSMMY-UHFFFAOYSA-N
SMILESFC1=CC=C(Cl)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4725-1.4765 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only