3,5-Difluoroanisole, 97%, Thermo Scientific Chemicals
3,5-Difluoroanisole, 97%, Thermo Scientific Chemicals
3,5-Difluoroanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Difluoroanisole, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A15497.14
also known as A15497-14
25 g
Catalog number A15497.14
also known as A15497-14
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS93343-10-3
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4630-1.4680 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only