Thermo Scientific Chemicals

Aminoacetaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A15498.22
100 g, Each
Thermo Scientific Chemicals

Aminoacetaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A15498.22
100 g, Each
Quantity
Catalog number: A15498.22
also known as A15498-22
Price (USD)
Price: 148.00
Online price: 125.65
Your price:
Quantity
-

Chemical Identifiers

CAS
22483-09-6
IUPAC Name
2,2-dimethoxyethan-1-amine
Molecular Formula
C4H11NO2
InChI Key
QKWWDTYDYOFRJL-UHFFFAOYSA-N
SMILES
COC(CN)OC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4155-1.4180 @ 20°C

Description

Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

Solubility
Miscible in water. Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only