Phthalide, 98+%
Phthalide, 98+%
Phthalide, 98+%
Thermo Scientific Chemicals

Phthalide, 98+%

CAS: 87-41-2 | C8H6O2 | 134.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A15516.36
also known as A15516-36
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS87-41-2
IUPAC Name1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC8H6O2
InChI KeyWNZQDUSMALZDQF-UHFFFAOYSA-N
SMILESO=C1OCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point70.5-76.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only