o-Benzoic sulfimide sodium salt hydrate, 99%
o-Benzoic sulfimide sodium salt hydrate, 99%
o-Benzoic sulfimide sodium salt hydrate, 99%
Thermo Scientific Chemicals

o-Benzoic sulfimide sodium salt hydrate, 99%

CAS: 82385-42-0 | C7H4NNaO3S | 205.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A15530.0I
also known as A15530-0I
Price (USD)
467.65
Online Exclusive
519.00
Save 51.35 (10%)
Each
Quantity:
5000 g
Request bulk or custom format
Price (USD)
467.65
Online Exclusive
519.00
Save 51.35 (10%)
Each
Chemical Identifiers
CAS82385-42-0
IUPAC Namesodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate
Molecular FormulaC7H4NNaO3S
InChI KeyOAZGZMKGRPRHJX-UHFFFAOYSA-M
SMILES[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤18%
Appearance (Color)White
O-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Effective catalyst for the silylation of a wide range of functional groups by Hexamethyl disilazane, A15139: J. Org. Chem., 47, 3966 (1982).
  2. Park, D. Y. Study of Stress Changes in Nanocrystalline Ni Thin Films Eletrodeposited from Chloride Baths. J. Korean electrochem. soc. 2011, 14 (3), 163-170.
  3. Zhao, S.; Yang, X.; Zhao, H.; Dong, A.; Wang, J.; Zhang, M.; Huang, W. Water-compatible surface imprinting of ‘Saccharin sodium’on silica surface for selective recognition and detection in aqueous solution. Talanta 2015, 144, 717-725.