Chlorotriethylsilane, 98+%
Chlorotriethylsilane, 98+%
Chlorotriethylsilane, 98+%
Thermo Scientific Chemicals

Chlorotriethylsilane, 98+%

CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15547.18
also known as A15547-18
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS994-30-9
IUPAC Namechlorotriethylsilane
Molecular FormulaC6H15ClSi
InChI KeyDCFKHNIGBAHNSS-UHFFFAOYSA-N
SMILESCC[Si](Cl)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0% (non-U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Appearance (Color)Clear colorless to pale yellow
CommentSpecification differs for U.S. and non-U.S. material where indicated
FormLiquid
View more
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

Solubility
Soluble in aprotic organic solvents.

Notes
Incompatible with bases, strong oxidizing agents, strong acids, alcohols, amines, ammonia, aldehydes and acetone.
RUO – Research Use Only

General References:

  1. Lakhtin, V. G.; Eremeeva, M. I.; Gordeev, E. G.; Ushakov, N. V.; Bykovchenko, V. G.; Kirilin, A. D.; Chernyshev, E. A. Reaction of chloro(ethyl)silanes with chloro(phenyl)silanes in the presence of aluminum chloride. Synthesis of chloro(ethyl)(phenyl)silanes. Russ. J. Gen. Chem. 2015, 85 (3), 595-599.
  2. Lu, H.; Hinkelmann, B.; Tautz, T.; Li, J.; Sasse, F.; Franke, R.; Kalesse, M. Paleo-soraphens: chemical total syntheses and biological studies. Org. Biomol. Chem. 2015, 13 (29), 8029-8036.