N-Methyl-4-nitroaniline, 97%, Thermo Scientific Chemicals
N-Methyl-4-nitroaniline, 97%, Thermo Scientific Chemicals
N-Methyl-4-nitroaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyl-4-nitroaniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15548.22
also known as A15548-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS100-15-2
IUPAC NameN-methyl-4-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyXIFJZJPMHNUGRA-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (GC)≥96.0%
Melting Point (clear melt)147.0-156.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only