Thermo Scientific Chemicals

Itaconic acid, 99%, Thermo Scientific Chemicals

Catalog number: A15566.30
250 g, Each
Thermo Scientific Chemicals

Itaconic acid, 99%, Thermo Scientific Chemicals

Catalog number: A15566.30
250 g, Each
Quantity
Catalog number: A15566.30
also known as A15566-30
Price (USD)
Price: 23.60
Online price: 20.65
Your price:
Quantity
-

Chemical Identifiers

CAS
97-65-4
IUPAC Name
2-methylidenebutanedioic acid
Molecular Formula
C5H6O4
InChI Key
LVHBHZANLOWSRM-UHFFFAOYSA-N
SMILES
OC(=O)CC(=C)C(O)=O
Appearance (Color)
White
Comment
Material Sourced in UK and US
Form
Crystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms (UK Sourced Material)

Description

Itaconic acid is used in the preparation of acrylonitrile-butadiene-styrene and acrylate latexes. It is also used to prepare poly-itaconic acid, resins biofuel components and ionomer cements. It finds application in the textile, chemical and pharmaceutical industries. It is also used as an additive in fibers and ion exchange resins to increase abrasion, waterproofing, physical resistance, dying affinity and better duration. Further, it acts as a co-monomer used in the preparation of acrylic fibers and rubbers, reinforced glass fiber, artificial diamonds and lens. In addition to this, it acts as a binder and sizing agent in non-weaving fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic acid is used in the preparation of acrylonitrile-butadiene-styrene and acrylate latexes. It is also used to prepare poly-itaconic acid, resins biofuel components and ionomer cements. It finds application in the textile, chemical and pharmaceutical industries. It is also used as an additive in fibers and ion exchange resins to increase abrasion, waterproofing, physical resistance, dying affinity and better duration. Further, it acts as a co-monomer used in the preparation of acrylic fibers and rubbers, reinforced glass fiber, artificial diamonds and lens. In addition to this, it acts as a binder and sizing agent in non-weaving fibers.

Solubility
Soluble in water, acetone, methanol, hexane and ethanol. Slightly soluble in benzene, chloroform, carbon disulfide and petroleum ether.

Notes
Hygroscopic. Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text