Allantoin, 98%
Allantoin, 98%
Allantoin, 98%
Thermo Scientific Chemicals

Allantoin, 98%

CAS: 97-59-6 | C4H6N4O3 | 158.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A15571.0B
also known as A15571-0B
Price (USD)
141.65
Special offer
Online exclusive
Ends: 28-Dec-2026
167.00
Save 25.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 28-Dec-2026
167.00
Save 25.35
Each
Chemical Identifiers
CAS97-59-6
IUPAC Name(2,5-dioxoimidazolidin-4-yl)urea
Molecular FormulaC4H6N4O3
InChI KeyPOJWUDADGALRAB-UHFFFAOYNA-N
SMILESNC(=O)NC1NC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
FormPowder
Identification (FTIR)Conforms
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

Solubility
Slightly soluble in water. Freely soluble in alkalis

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. Sevgi Yardim-Akaydin; Aylin Sepici; Yeşim Ozkan; Meral Torun; Bolkan Simşek; Vesile Sepici. Oxidation of uric acid in rheumatoid arthritis: is allantoin a marker of oxidative stress?. Free Radical Research. 2004,38 (6), 623-628.
  2. A V Shestopalov; T P Shkurat; Z I Mikashinovich; I O Kryzhanovskaia; M A Bogacheva; S V Lomteva; V P Prokof'ev; E P Gus'kov. [Biological functions of allantoin]. Izvestiia Akademii nauk. Seriia biologicheskaia / Rossiiskaia akademiia nauk. 2006, 5, 541-545.