Would I be able to find out the sex of our Primary Mouse Cortical Neurons?
Primary Mouse Cortical Neurons are mixed sex, because it's too hard to identify the sex of a mouse fetus.
Search
View additional product information for Primary Mouse Cortical Neurons - FAQs (A15586, A15585)
3 product FAQs found