Ethyl 2,3-dibromopropionate, 97%
Ethyl 2,3-dibromopropionate, 97%
Ethyl 2,3-dibromopropionate, 97%
Thermo Scientific Chemicals

Ethyl 2,3-dibromopropionate, 97%

CAS: 3674-13-3 | C5H8Br2O2 | 259.925 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15591.22
also known as A15591-22
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS3674-13-3
IUPAC Nameethyl 2,3-dibromopropanoate
Molecular FormulaC5H8Br2O2
InChI KeyOENICUBCLXKLJQ-UHFFFAOYNA-N
SMILESCCOC(=O)C(Br)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4985-1.5035 @ 20?C
Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, bases.
RUO – Research Use Only

General References:

  1. Richard J. Jablonski; Eugene I. Snyder. Conformational stability in ethyl 2,3-dibromopropionate. J. Am. Chem. soc. 1968,90 (9), 2316-2318.
  2. Janina Karolak-Wojciechowska; Katarzyna Kieć-Kononowicz. Reaction of 5,5-diphenyl-2-thiohydantoin with ethyl-2,3-dibromopropionate. Crystal and molecular structure of ethyl-2-{2,3-dihydro-6,6-diphenylimidazo-[2,1-b]-thiazol-5(6H)-one} formate. Journal of Crystallographic and Spectroscopic Research. 1987, 17 (4), 485-494.