4-Picoline, 98%
4-Picoline, 98%
4-Picoline, 98%
Thermo Scientific Chemicals

4-Picoline, 98%

CAS: 108-89-4 | C6H7N | 93.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A15595.AP
also known as A15595-AP
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.90
Save 7.25 (15%)
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.90
Save 7.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS108-89-4
IUPAC Name4-methylpyridine
Molecular FormulaC6H7N
InChI KeyFKNQCJSGGFJEIZ-UHFFFAOYSA-N
SMILESCC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay from Supplier's CofA≥97.5% (US Sourced Material)
Identification (FTIR)Conforms (UK Sourced Material)
CommentMaterial Sourced in UK and US
Assay (GC)≥97.5% (UK Sourced Material)
View more
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

Solubility
Soluble in water, alcohol and ether.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
RUO – Research Use Only

General References:

  1. Zafar A. Siddiqi; Mohd Khalid; Sarvendra Kumar; M. Shahid; Shabana Noor. Antimicrobial and SOD activities of novel transition metal complexes of pyridine-2,6-dicarboxylic acid containing 4-picoline as auxiliary ligand. European Journal of Medicinal Chemistry. 2010,45 (1), 264-269.
  2. Takeshi Sakamoto; John M. Joern; Akira Arisawa; and Frances H. Arnold. Laboratory Evolution of Toluene Dioxygenase To Accept 4-Picoline as a Substrate. Appl. Environ. Microbiol. 2001, 67 (9), 3882-3887.