4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.
Solubility
Soluble in alcohol, ethanol (0.5g/10 mL). Insoluble in water.
Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- K Aoki; and K Koga. Stereoselective reactions. XXXII. Enantioselective deprotonation of 4-tert-butylcyclohexanone by fluorine-containing chiral lithium amides derived from 1-phenylethylamine and 1-(1-naphthyl)ethylamine. Chemical & Pharmaceutical Bulletin. 2000, 48 (4), 571-574.
- John T. Edward; Stereochemistry of the Bucherer-Bergs and Strecker Reactions of 4-tert-Butylcyclohexanone. Canadian Journal of Chemistry. 1975, 53( (22), 3339-3350.