6-Chloronicotinic acid, 99%
6-Chloronicotinic acid, 99%
6-Chloronicotinic acid, 99%
6-Chloronicotinic acid, 99%
Thermo Scientific Chemicals

6-Chloronicotinic acid, 99%

CAS: 5326-23-8 | C6H4ClNO2 | 157.553 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15620.06
also known as A15620-06
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Add to cart
Chemical Identifiers
CAS5326-23-8
IUPAC Name6-chloropyridine-3-carboxylic acid
Molecular FormulaC6H4ClNO2
InChI KeyUAWMVMPAYRWUFX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormPowder
Assay (Silylated GC)≥98.5%
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
6-Chloropyridine-3-carboxylic acid (6-chloronicotinic acid/6-CNA) has been used to study its photolytic and photocatalytic degradation. The product has been used as a media component during the isolation of 6-CNA degrading bacterial strain from imidacloprid-exposed soil samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropyridine-3-carboxylic acid (6-chloronicotinic acid/6-CNA) has been used to study its photolytic and photocatalytic degradation. The product has been used as a media component during the isolation of 6-CNA degrading bacterial strain from imidacloprid-exposed soil samples.

Solubility
Soluble in water (2 mg/ml at 20°C), ethanol, methanol, DMSO, and slightly soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Romina Žabar, et al. Photolytic and photocatalytic degradation of 6-chloronicotinic acid.Chemosphere.,2011,85(5), 861-868.
  2. Madhura Shettigar, et al. Cloning of a novel 6-chloronicotinic acid chlorohydrolase from the newly isolated 6-chloronicotinic acid mineralizing Bradyrhizobiaceae strain SG-6C.PLoS One.,2012,7(11), 51162.