Sodium tellurite(IV), 97%
Sodium tellurite(IV), 97%
Sodium tellurite(IV), 97%
Thermo Scientific Chemicals

Sodium tellurite(IV), 97%

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A15633.18
also known as A15633-18
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Specifications
CAS10102-20-2
Chemical Name or MaterialSodium tellurite(IV)
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.
EINECS Number233-268-4
Formula Weight221.58
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only