3-Fluorophenylacetic acid, 98%
3-Fluorophenylacetic acid, 98%
3-Fluorophenylacetic acid, 98%
3-Fluorophenylacetic acid, 98%
Thermo Scientific Chemicals

3-Fluorophenylacetic acid, 98%

CAS: 331-25-9 | C8H7FO2 | 154.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15635.22
also known as A15635-22
Price (USD)
470.65
Online Exclusive
523.00
Save 52.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
470.65
Online Exclusive
523.00
Save 52.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS331-25-9
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or flakes
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Appearance (Color)White to pale cream
Water Content (Karl Fischer Titration)≤1.5%
View more
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

Solubility
Slightly soluble in chloroform and methanol. Insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Adel Nefzi; Jon Appel; Sergey Arutyunyan; Richard A Houghten; Adel Nefzi; Jon Appel; Sergey Arutyunyan; Richard A. Houghten. Parallel synthesis of chiral pentaamines and pyrrolidine containing bis-heterocyclic libraries. Multiple scaffolds with multiple building blocks: A double diversity for the identification of new antitubercular compounds. Bioorganic & Medicinal Chemistry Letters. 2009, 19 (17), 5169-5175.
  2. Marisa Cabeza; Eugene Bratoeff; Georgina Gómez; Ivonne Heuze; Arely Rojas; Martha Ochoa; Miguel Angel Palomino; Cristina Revilla. Synthesis and biological effect of halogen substituted phenyl acetic acid derivatives of progesterone as potent progesterone receptor antagonists. The Journal of Steroid Biochemistry and Molecular Biology. 2008, 111 (3-5), 232-239.